missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-1H-indole-7-carboxylic acid, 97%, Thermo Scientific™

Catalog Number p-3586607
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
167479-16-5
C10H9NO2
175.19
MFCD09879957
DWKIPLIDZYRILV-UHFFFAOYSA-N
18388362
1-methylindole-7-carboxylic acid
CN1C=CC2=CC=CC(C(O)=O)=C12
This item is not returnable. View return policy
167479-16-5
C10H9NO2
175.19
MFCD09879957
DWKIPLIDZYRILV-UHFFFAOYSA-N
18388362
1-methylindole-7-carboxylic acid
CN1C=CC2=CC=CC(C(O)=O)=C12
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Synonym 1-methyl-1h-indole-7-carboxylic acid, 1-methyl-7-indolecarboxylic acid, 1h-indole-7-carboxylicacid, 1-methyl, 1-methyl-7-indolecarboxylicacid, 1h-indole-7-carboxylicacid,1-methyl, 1h-indole-7-carboxylic acid, 1-methyl
Formula Weight 175.19
Percent Purity 97%
Chemical Name or Material 1-Methyl-1H-indole-7-carboxylic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'