missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-1H-indole-6-carbaldehyde, 97%, Thermo Scientific™

Catalog Number p-4511343
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
21005-45-8
C10H9NO
159.19
MFCD08690251
QGAKKGIWURKKHV-UHFFFAOYSA-N
1-methyl-1h-indole-6-carbaldehyde, 1h-indole-6-carboxaldehyde, 1-methyl, 1h-indole-6-carboxaldehyde,1-methyl, 6-formy-1-methylindole, 6-formyl-1-methylindole, 1-methyl-1h-indole-6-carboxaldehyde
14835679
1-methylindole-6-carbaldehyde
CN1C=CC2=CC=C(C=O)C=C12
This item is not returnable. View return policy
21005-45-8
C10H9NO
159.19
MFCD08690251
QGAKKGIWURKKHV-UHFFFAOYSA-N
1-methyl-1h-indole-6-carbaldehyde, 1h-indole-6-carboxaldehyde, 1-methyl, 1h-indole-6-carboxaldehyde,1-methyl, 6-formy-1-methylindole, 6-formyl-1-methylindole, 1-methyl-1h-indole-6-carboxaldehyde
14835679
1-methylindole-6-carbaldehyde
CN1C=CC2=CC=C(C=O)C=C12
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 159.19
Percent Purity 97%
Chemical Name or Material 1-Methyl-1H-indole-6-carbaldehyde
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'