missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-1H-indole-4-sulfonyl chloride, 97%, Thermo Scientific™

Catalog Number p-3484805
Click to view available options
Quantity:
250 mg
1 g
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
876316-36-8
C9H8ClNO2S
229.678
MFCD08690260
HFWFOJFQXZBLAU-UHFFFAOYSA-N
1-methyl-1h-indole-4-sulfonyl chloride, 1h-indole-4-sulfonylchloride, 1-methyl, 1-methyl-1h-indole-4-sulphonyl chloride, chloro 1-methylindol-4-yl sulfone, 1-methyl-1h-indole-4-sulfonylchloride, 4-chlorosulphonyl-1-methyl-1h-indole, 1h-indole-4-sulfonyl chloride, 1-methyl
18525782
1-methylindole-4-sulfonyl chloride
CN1C=CC2=C1C=CC=C2S(=O)(=O)Cl
This item is not returnable. View return policy
876316-36-8
C9H8ClNO2S
229.678
MFCD08690260
HFWFOJFQXZBLAU-UHFFFAOYSA-N
1-methyl-1h-indole-4-sulfonyl chloride, 1h-indole-4-sulfonylchloride, 1-methyl, 1-methyl-1h-indole-4-sulphonyl chloride, chloro 1-methylindol-4-yl sulfone, 1-methyl-1h-indole-4-sulfonylchloride, 4-chlorosulphonyl-1-methyl-1h-indole, 1h-indole-4-sulfonyl chloride, 1-methyl
18525782
1-methylindole-4-sulfonyl chloride
CN1C=CC2=C1C=CC=C2S(=O)(=O)Cl
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 229.69
Percent Purity 97%
Chemical Name or Material 1-Methyl-1H-indole-4-sulfonyl chloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'