missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-methyl-1H-indole-4-carbonitrile, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Specifications
Assay Percent Range | 97% |
Quantity | 250 mg |
Synonym | 1-methyl-1h-indole-4-carbonitrile, 1h-indole-4-carbonitrile,1-methyl, 1h-indole-4-carbonitrile, 1-methyl |
IUPAC Name | 1-methylindole-4-carbonitrile |
Formula Weight | 156.19 |
Percent Purity | 97% |
Chemical Name or Material | 1-Methyl-1H-indole-4-carbonitrile |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'