missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-1H-indazole-3-carbaldehyde, 97%, Thermo Scientific™

Catalog Number p-3175468
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
4002-83-9
C9H8N2O
160.18
MFCD08060478
KYDLGYCLCFKUHY-UHFFFAOYSA-N
1-methyl-1h-indazole-3-carbaldehyde, 1-methyl-1h-indazole-3-carboxaldehyde, 3-formyl-1-methyl-1h-indazole, 1-methyl-3-formylindazole, 1h-indazole-3-carboxaldehyde,1-methyl
7164545
1-methylindazole-3-carbaldehyde
CN1N=C(C=O)C2=CC=CC=C12
This item is not returnable. View return policy
4002-83-9
C9H8N2O
160.18
MFCD08060478
KYDLGYCLCFKUHY-UHFFFAOYSA-N
1-methyl-1h-indazole-3-carbaldehyde, 1-methyl-1h-indazole-3-carboxaldehyde, 3-formyl-1-methyl-1h-indazole, 1-methyl-3-formylindazole, 1h-indazole-3-carboxaldehyde,1-methyl
7164545
1-methylindazole-3-carbaldehyde
CN1N=C(C=O)C2=CC=CC=C12
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 160.18
Percent Purity 97%
Chemical Name or Material 1-Methyl-1H-indazole-3-carbaldehyde
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'