missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-1H-benzotriazole, TRC

Product Code. 30471127
Change view
Click to view available options
Mængde:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30471127 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30471127 Supplier Toronto Research Chemicals Supplier No. TRCM2898101G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale 1-Methyl-1H-benzotriazole
CAS 13351-73-0
Molekylær formel C7 H7 N3
Mængde 1 g
Synonym 1-Methyl-1H-benzotriazole, 1-Methyl-1,2,3-benzotriazole, 1-Methyl-1H-1,2,3-benzotriazole, 1-Methyl-1H-benzo[d][1,2,3]triazole, 1-Methylbenzotriazole, NSC 11743, 1H-Benzotriazole, 1-methyl-, 1-Methyl-1,2,3-benzotriazole, 1-Methylbenzotriazole, NSC 11743, 1-Methyl-1H-benzotriazole, 1-Methyl-1,2,3-benzotriazole, 1-Methyl-1H-1,2,3-benzotriazole, 1-Methyl-1H-benzo[d][1,2,3]triazole, 1-Methylbenzotriazole, NSC 11743, 1H-Benzotriazole, 1-methyl-
InChI formel InChI=1S/C7H7N3/c1-10-7-5-3-2-4-6(7)8-9-10/h2-5H,1H3
SMIL Cn1nnc2ccccc12
IUPAC navn 1-methylbenzotriazole
Molekylvægt (g/mol) 133.15
Formel vægt 133.064
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.