Få mere at vide
1-Methyl-12-thioxopyrido[1,2,3,4]imidazole-[1,2-a]benzimidazole-2(12H)-one, TRC
Artikelnummer. 30476674
Skift visning
Mængde:
5 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30476674 100 mg
30482325 25 mg
30476871 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30476674 Leverandør Toronto Research Chemicals Leverandørnr. TRCM330550100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1246819-06-6
Molekylær formel C14 H9 N3 O S
Molekylvægt (g/mol) 267.31
Synonym 1-Methyl-12-thioxopyrido[1',2':3,4]imidazo[1,2-a]benzimidazol-2(12H)-one
IUPAC navn 1-Methyl-12-thioxopyrido[1',2':3,4]imidazo[1,2-a]benzimidazol-2(12H)-one
SMIL CC1=C2N(C=CC1=O)c3nc4ccccc4n3C2=S
Tekniske data
Kemisk navn eller materiale 1-Methyl-12-thioxopyrido[1',2':3,4]imidazo[1,2-a]benzimidazol-2(12H)-one
Mængde 100 mg
InChI formel InChI=1S/C14H9N3OS/c1-8-11(18)6-7-16-12(8)13(19)17-10-5-3-2-4-9(10)15-14(16)17/h2-7H,1H3
Formel vægt 267.0466