Få mere at vide
1-Methyl-1,4-dihydronicotinamide, TRC
Artikelnummer. 30138083
Skift visning
Mængde:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30138083 100 mg
30137742 500 mg
30011844 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30138083 Leverandør Toronto Research Chemicals Leverandørnr. TRCM302175100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 17750-23-1
Molekylær formel C7 H10 N2 O
Molekylvægt (g/mol) 138.17
IUPAC navn 1-methyl-4H-pyridine-3-carboxamide
SMIL CN1C=CCC(=C1)C(=O)N
Tekniske data
Kemisk navn eller materiale 1-Methyl-1,4-dihydronicotinamide
Mængde 100 mg
InChI formel InChI=1S/C7H10N2O/c1-9-4-2-3-6(5-9)7(8)10/h2,4-5H,3H2,1H3,(H2,8,10)
Formel vægt 138.0793