Learn More
1-(Mercaptomethyl)cyclopropaneacetic Acid (2-[1-(Sulfanylmethyl)cyclopropyl]acetic Acid), Mikromol™
Product Code. 30049948
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Product Code. Quantity Unit Size
30049948 25 mg 25mg
1 options
Product Code. 30049948 Supplier Mikromol Supplier No. MM0698.230025
 Restricted Item
Forventet forsendelse: 30-11-2026
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
1-(Mercaptomethyl)cyclopropaneacetic Acid (2-[1-(Sulfanylmethyl)cyclopropyl]acetic Acid)
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 1-(Mercaptomethyl)cyclopropaneacetic Acid
CAS 162515-68-6
Analyte or Component Names 1-(Mercaptomethyl)cyclopropaneacetic Acid
Shipping Condition Room Temperature
Molecular Formula C6 H10 O2 S
Certifications/Compliance ISO 9001
Synonym 1-(Mercaptomethyl)cyclopropaneacetic Acid, [1-(Mercaptomethyl)cyclopropyl]acetic Acid
InChI Formula InChI=1S/C6H10O2S/c7-5(8)3-6(4-9)1-2-6/h9H,1-4H2,(H,7,8)
SMILES OC(=O)CC1(CS)CC1
IUPAC Name 2-[1-(sulfanylmethyl)cyclopropyl]acetic acid
Molecular Weight (g/mol) 146.21
Quantity 25 mg
Formula Weight 146.0402 g/mol