Få mere at vide
1-Linoleoyl-(2S)-glycerol, TRC
Artikelnummer. 30477293
Skift visning
Mængde:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30477293 50 mg
30471428 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30477293 Leverandør Toronto Research Chemicals Leverandørnr. TRCL46795550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 67968-46-1
Molekylær formel C21H38O4
Molekylvægt (g/mol) 354.52
Synonym α-Linoleic Acid (2S)-2,3-Dihydroxypropyl Ester, (9Z,12Z)-9,12-Octadecadienoic Acid (2S)-2,3-Dihydroxypropyl Ester
IUPAC navn [(2S)-2,3-dihydroxypropyl] (9Z,12Z)-octadeca-9,12-dienoate
SMIL CCCCC/C=C\C/C=C\CCCCCCCC(=O)OC[C@H](CO)O
Tekniske data
Kemisk navn eller materiale 1-Linoleoyl-(2S)-glycerol
Mængde 50 mg
InChI formel InChI=1S/C21H38O4/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-21(24)25-19-20(23)18-22/h6-7,9-10,20,22-23H,2-5,8,11-19H2,1H3/b7-6-,10-9-/t20-/m0/s1
Formel vægt 354.277