Få mere at vide
1-Keto Bambuterol Hydrochloride, TRC
Artikelnummer. 30408010
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30408010 25 mg
1 options
Artikelnummer. 30408010 Leverandør Toronto Research Chemicals Leverandørnr. TRCK17150025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1-Keto Bambuterol Hydrochloride
CAS 2153431-91-3
Molekylær formel C18H27N3O5 . HCl
Mængde 25 mg
Synonym N,N-Dimethylcarbamic Acid C,C'-[5-[[(1,1-Dimethylethyl)amino]acetyl]-1,3-phenylene] Ester Hydrochloride, Dimethylcarbamic Acid 5-[[(1,1-Dimethylethyl)amino]acetyl]-1,3-phenylene Ester Hydrochloride, Bis[3',5'-(N,N-dimethylcarbamoyloxi)]-2-N-tert-butylaminoacetophenone Hydrochloride
InChI formel InChI=1S/C18H27N3O5.ClH/c1-18(2,3)19-11-15(22)12-8-13(25-16(23)20(4)5)10-14(9-12)26-17(24)21(6)7;/h8-10,19H,11H2,1-7H3;1H
SMIL Cl.CN(C)C(=O)Oc1cc(OC(=O)N(C)C)cc(c1)C(=O)CNC(C)(C)C
IUPAC navn [3-[2-(tert-butylamino)acetyl]-5-(dimethylcarbamoyloxy)phenyl] N,N-dimethylcarbamate;hydrochloride
Molekylvægt (g/mol) 401.9
Formel vægt 401.1717