missing translation for 'onlineSavingsMsg'
Learn More

1-Isopropylpiperazine, 98%, Thermo Scientific Chemicals

Catalog Number p-7028169
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
4318-42-7
C7H16N2
128.219
MFCD00167971
WHKWMTXTYKVFLK-UHFFFAOYSA-N
1-isopropylpiperazine, n-isopropylpiperazine, 1-propan-2-yl piperazine, piperazine, 1-isopropyl, isopropyl piperazine, 1-isopropyl-piperazine, 1-1-methylethyl piperazine, n-isopropyl piperazine, 1-isopropyl piperazine, piperazine, 1-1-methylethyl
78013
1-propan-2-ylpiperazine
CC(C)N1CCNCC1
This item is not returnable. View return policy
4318-42-7
C7H16N2
128.219
MFCD00167971
WHKWMTXTYKVFLK-UHFFFAOYSA-N
1-isopropylpiperazine, n-isopropylpiperazine, 1-propan-2-yl piperazine, piperazine, 1-isopropyl, isopropyl piperazine, 1-isopropyl-piperazine, 1-1-methylethyl piperazine, n-isopropyl piperazine, 1-isopropyl piperazine, piperazine, 1-1-methylethyl
78013
1-propan-2-ylpiperazine
CC(C)N1CCNCC1

1-Isopropylpiperazine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Isopropylpiperazine is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Density 0.896
Boiling Point 180°C to 181°C
Flash Point 54°C (130°F)
Refractive Index 1.47
Quantity 1 g
UN Number UN2733
Solubility Information Slightly soluble in water.
Formula Weight 128.22
Percent Purity 98%
Chemical Name or Material 1-Isopropylpiperazine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'