Få mere at vide
1-Isopropylamino-3-phenoxypropan-2-ol, TRC
Artikelnummer. 17993275
Skift visning
Mængde:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17993275 1 g
17903175 5 g
17952865 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 17993275 Leverandør Toronto Research Chemicals Leverandørnr. TRCI8240701G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 7695-63-8
Molekylær formel C12 H19 N O2
Molekylvægt (g/mol) 209.28
Synonym (2RS)-1-[(1-Methylethyl)amino]-3-phenoxypropan-2-ol, Metoprolol Succinate Imp. F (EP), Metoprolol Tartrate Imp. F (EP)
IUPAC navn 1-phenoxy-3-(propan-2-ylamino)propan-2-ol
SMIL CC(C)NCC(O)COc1ccccc1
Tekniske data
Kemisk navn eller materiale (2RS)-1-[(1-Methylethyl)amino]-3-phenoxy-propan-2-ol
Mængde 1 g
InChI formel InChI=1S/C12H19NO2/c1-10(2)13-8-11(14)9-15-12-6-4-3-5-7-12/h3-7,10-11,13-14H,8-9H2,1-2H3
Formel vægt 209.1416