Få mere at vide
1-Iodopropane-d7, CDN
Artikelnummer. 30320487
Skift visning
Mængde:
0.5 g
1 g
Pakningsstørrelse:
0.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30320487 1 g -
30318942 0.5 g 0.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30320487 Leverandør CDN Leverandørnr. CDND12371G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1-Iodopropane-d7
CAS 59012-23-6
Molekylær formel C3 D7 I
Mængde 1 g
Synonym Propane-1,1,1,2,2,3,3-d7, 3-iodo- (9 CI), 1-Iodopropane-d7, 3-Iodopropane-1,1,1,2,2,3,3-d7, 1-Iodopropane-1,1,2,2,3,3,3-d7, 1-Iodopropane-d7, 1-Iodopropane-D7, Propyl-d7 iodide
InChI formel InChI=1 S/C3H7I/c1-2-3-4/h2-3H2,1H3/i1D3,2D2,3D2
SMIL [2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])I
IUPAC navn 1,1,1,2,2,3,3-heptadeuterio-3-iodopropane
Molekylvægt (g/mol) 177.0353
Formel vægt 177.0032 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity