Få mere at vide
1-Iodopropane-1,1,2,2-d4, CDN
Artikelnummer. 30315522
Skift visning
Mængde:
0.1 g
0.25 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30315522 0.25 g -
30310983 0.1 g 0.1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30315522 Leverandør CDN Leverandørnr. CDND42280.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1-Iodopropane-1,1,2,2-D4
CAS 1219804-45-1
Molekylær formel C3 D4 H3 I
Mængde 0.25 g
Synonym 1-Iodopropane-1,1,2,2-d4, 1-Iodopropane-d4, 1-Iodopropane-1,1,2,2-D4, [1,1,2,2-2H4]Propyl iodide
InChI formel InChI=1 S/C3H7I/c1-2-3-4/h2-3H2,1H3/i2D2,3D2
SMIL [2 H]C([2 H])(C)C([2 H])([2 H])I
IUPAC navn 1,1,2,2-tetradeuterio-1-iodopropane
Molekylvægt (g/mol) 174.0168
Formel vægt 173.9844 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity