Få mere at vide
1-Iodooctane-d17, CDN
Artikelnummer. 30316765
Skift visning
Mængde:
0.25 g
0.5 g
Pakningsstørrelse:
0.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30316765 0.5 g 0.5g
30310855 0.25 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30316765 Leverandør CDN Leverandørnr. CDND12350.5G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1-Iodooctane-D17
CAS 340256-37-3
Molekylær formel C8 D17 I
Mængde 0.5 g
Synonym Octane-1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8-d17, 8-iodo- (9 CI), 8-Iodooctane-1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8-d17, 8-Iodooctane-d17, 1-Iodooctane-d17, 1-Iodooctane-D17
InChI formel InChI=1 S/C8H17I/c1-2-3-4-5-6-7-8-9/h2-8H2,1H3/i1D3,2D2,3D2,4D2,5D2,6D2,7D2,8D2
SMIL [2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])I
IUPAC navn 1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8-heptadecadeuterio-8-iodooctane
Molekylvægt (g/mol) 257.2298
Formel vægt 257.1442 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity