missing translation for 'onlineSavingsMsg'
Learn More

1-Iodohexane, 98%, stabilized, ACROS Organics™

Brand:  Acros Organics 218525000

Additional Details : CAS Number : 638-45-9 Weight : 0.77000kg

Packaging Glass bottle
Quantity 500mL

 View more versions of this product

Product Code. 10209733

  • 1568.00 DKK / 500mL

In stock, ready to ship: 1

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

638-45-9
212.07
ANOOTOPTCJRUPK-UHFFFAOYSA-N
12527
CCCCCCI
C6H13I
MFCD00001102
hexyl iodide, n-hexyl iodide, hexane, 1-iodo, 1-hexyl iodide, iodohexane, 1-iodo-hexane, hexane,1-iodo, n-hexyliodide, iodo-hexane, 1-iodanylhexane
1-iodohexane

Specifications

1-Iodohexane
61°C
Glass bottle
1.437
179.0°C to 180.0°C
500mL
638-45-9
100.0
C6H13I
MFCD00001102
hexyl iodide, n-hexyl iodide, hexane, 1-iodo, 1-hexyl iodide, iodohexane, 1-iodo-hexane, hexane,1-iodo, n-hexyliodide, iodo-hexane, 1-iodanylhexane
ANOOTOPTCJRUPK-UHFFFAOYSA-N
1-iodohexane
12527
Liquid
1.4370g/mL
Authentic
1.4910 to 1.4930
copper
Colorless to Yellow
Stabilized
98.0
97.5% min. (GC)
CH3(CH2)5I
01, 146
Solubility in water: insoluble
CCCCCCI
212.07
212.07
98%
Videos
SDS Documents