Få mere at vide
1-Iodobutane-d9, CDN
Artikelnummer. 30323069
Skift visning
Mængde:
1 g
Artikelnummer. Mængde
30323069 1 g
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30323069 Leverandør CDN Leverandørnr. CDND12301G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1-Iodobutane-d9
CAS 59012-24-7
Molekylær formel C4 D9 I
Mængde 1 g
Synonym Butane-1,1,1,2,2,3,3,4,4-d9, 4-iodo- (9 CI), 1-Iodobutane-d9, Perdeuteriobutyl iodide, 4-Iodobutane-1,1,1,2,2,3,3,4,4-d9, 1-Iodobutane-D9, Butyl-d9 iodide
InChI formel InChI=1 S/C4H9I/c1-2-3-4-5/h2-4H2,1H3/i1D3,2D2,3D2,4D2
SMIL [2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])I
IUPAC navn 1,1,1,2,2,3,3,4,4-nonadeuterio-4-iodobutane
Molekylvægt (g/mol) 193.074
Formel vægt 193.031 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity