Få mere at vide
1-Hydroxyadamantane-d15, TRC
Artikelnummer. 30322037
Skift visning
Mængde:
1 mg
5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30322037 1 mg
30320136 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30322037 Leverandør Toronto Research Chemicals Leverandørnr. TRCA2079111MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1-Adamantanol-D15
CAS 33830-11-4
Molekylær formel C10 D15 H O
Mængde 1 mg
Synonym 1-Adamantan-d15-ol (8CI), 1-Hydroxyadamantane-d15, 1-Adamantanol-D15
InChI formel InChI=1S/C10H16O/c11-10-4-7-1-8(5-10)3-9(2-7)6-10/h7-9,11H,1-6H2/i1D2,2D2,3D2,4D2,5D2,6D2,7D,8D,9D
SMIL [2H]C1([2H])C2([2H])C([2H])([2H])C3([2H])C([2H])([2H])C1([2H])C([2H])([2H])C(O)(C3([2H])[2H])C2([2H])[2H]
IUPAC navn 2,2,3,4,4,5,6,6,7,8,8,9,9,10,10-pentadecadeuterioadamantan-1-ol
Molekylvægt (g/mol) 167.3259
Formel vægt 167.2143