missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Heptanol, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
Description
1-Heptanol, is used in the fragrance industry due to its pleasant smell.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Heptanol, is used in the fragrance industry due to its pleasant smell.
Solubility
Soluble in water 1.7 g/L at 25°C.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.Stable under recommended storage conditions. Incompatible to strong oxidizing agents, acid chlorides, acid anhydrides.
1-Heptanol, is used in the fragrance industry due to its pleasant smell.
Solubility
Soluble in water 1.7 g/L at 25°C.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.Stable under recommended storage conditions. Incompatible to strong oxidizing agents, acid chlorides, acid anhydrides.
Specifications
Melting Point | -34°C |
Density | 0.82 |
Boiling Point | 175°C to 176°C |
Flash Point | 73°C (163°F) |
Assay Percent Range | 99% |
Odor | Characteristic |
Linear Formula | CH3(CH2)6OH |
Refractive Index | 1.424 |
Quantity | 100 mL |
Beilstein | 1731686 |
Show More |
Safety and Handling
Product Identifier
- 1-Heptanol
- Warning
- Serious eye damage/eye irritation Category 2
- H319-Causes serious eye irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- MIXTURE LIST-Contains : 1-Heptanol
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'