Få mere at vide
1-Formyl-4-(2,3,4-trimethoxybenzyl)piperazine Hydrochloride, Mikromol™
Artikelnummer. 17970796
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17970796 25 mg
17902906 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 17970796 Leverandør Mikromol Leverandørnr. MM0539.080025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
1-Formyl-4-(2,3,4-trimethoxybenzyl)piperazine Hydrochloride
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1-Formyl-4-(2,3,4-trimethoxybenzyl)piperazine Hydrochloride
CAS 879646-17-0
Analyt- eller komponentnavne 1-Formyl-4-(2,3,4-trimethoxybenzyl)piperazine Hydrochloride
Forsendelsestilstand Room Temperature
Molekylær formel C15 H22 N2 O4 . Cl H
Certificeringer/compliance ISO 17025
Synonym 1-Formyl-4-(2,3,4-trimethoxybenzyl)piperazine Hydrochloride
InChI formel InChI=1S/C15H22N2O4.ClH/c1-19-13-5-4-12(14(20-2)15(13)21-3)10-16-6-8-17(11-18)9-7-16;/h4-5,11H,6-10H2,1-3H3;1H
SMIL Cl.COc1ccc(CN2CCN(CC2)C=O)c(OC)c1OC
IUPAC navn 4-[(2,3,4-trimethoxyphenyl)methyl]piperazine-1-carbaldehyde;hydrochloride
Molekylvægt (g/mol) 330.81
Mængde 25 mg
Formel vægt 330.1346 g/mol