Learn More
Alfa Aesar™ 1-(Ethylsulfonyl)piperazine, 97%
Brand: Alfa Aesar™ H55503.03
Additional Details : CAS Number : 62937-96-6 Weight : 0.00100kg
Quantity | 1g |
---|
Chemical Identifiers
62937-96-6 | |
178.25 | |
BIYGAOBOLDXNHM-UHFFFAOYSA-N | |
415294 | |
CCS(=O)(=O)N1CCNCC1 |
C6H14N2O2S | |
MFCD02093528 | |
1-ethylsulfonyl piperazine, 1-ethylsulphonyl piperazine, 1-ethanesulfonyl piperazine, 1-ethylsulfonyl-piperazine, 1-ethanesulfonyl-piperazine, 1-ethanesulfonylpiperazine, ethylsulfonyl piperazine, ethanesulfonylpiperazine, ethanesulfonyl-piperazine | |
1-ethylsulfonylpiperazine |
Specifications
1-(Ethylsulfonyl)piperazine | |
69°C to 72°C | |
62937-96-6 | |
MFCD02093528 | |
BIYGAOBOLDXNHM-UHFFFAOYSA-N | |
1-ethylsulfonylpiperazine | |
415294 | |
Solid |
Brown to White | |
1g | |
C6H14N2O2S | |
1-ethylsulfonyl piperazine, 1-ethylsulphonyl piperazine, 1-ethanesulfonyl piperazine, 1-ethylsulfonyl-piperazine, 1-ethanesulfonyl-piperazine, 1-ethanesulfonylpiperazine, ethylsulfonyl piperazine, ethanesulfonylpiperazine, ethanesulfonyl-piperazine | |
CCS(=O)(=O)N1CCNCC1 | |
178.25 | |
178.25 | |
97% |