Få mere at vide
1-Ethyldihydro-6-imino-2,4,5(3H)-pyrimidinetrione 5-(O-Methyloxime), TRC
Artikelnummer. 30137200
Skift visning
Mængde:
50 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30137200 250 mg
30010662 50 mg
30005463 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30137200 Leverandør Toronto Research Chemicals Leverandørnr. TRCE817925250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 71342-66-0
Molekylær formel C7H10N4O3
Molekylvægt (g/mol) 198.18
SMIL O=Cc1nc(C)oc1
Tekniske data
Kemisk navn eller materiale 1-Ethyldihydro-6-imino-2,4,5(3H)-pyrimidinetrione 5-(O-Methyloxime)
Mængde 250 mg
InChI formel InChI=1S/C7H10N4O3/c1-3-11-5(8)4(10-14-2)6(12)9-7(11)13/h8H,3H2,1-2H3,(H,9,12,13)/b8-5?,10-4-