Få mere at vide
1-Ethyl-4-nitrosopiperazine, TRC
Artikelnummer. 30371951
Skift visning
Mængde:
50 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30371951 500 mg
30369351 250 mg
30371273 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30371951 Leverandør Toronto Research Chemicals Leverandørnr. TRCE928435500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 65504-33-8
Molekylær formel C6H13N3O
Molekylvægt (g/mol) 143.19
Synonym Piperazine, 1-ethyl-4-nitroso- (9CI, ACI), 1-Ethyl-4-nitrosopiperazine
SMIL CCN1CCN(CC1)N=O
Tekniske data
Kemisk navn eller materiale 1-Ethyl-4-nitrosopiperazine
Mængde 500 mg
InChI formel InChI=1S/C6H13N3O/c1-2-8-3-5-9(7-10)6-4-8/h2-6H2,1H3
Formel vægt 143.1059