missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-Ethyl-4-(2-fluoro-4-nitrophenyl)piperazine, 97%

Brand:  Alfa Aesar™ H63843.03

Additional Details : CAS Number : 694501-34-3 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15470759

  • 704.62 DKK / 1g


Estimated Shipment date
from Supplier 15-01-2019
Add to basket

Specifications

Chemical Identifiers

694501-34-3
253.277
1-ethyl-4-2-fluoro-4-nitrophenyl piperazine, 1-ethyl-4-2-fluoro-4-nitro-phenyl-piperazine, piperazine, 1-ethyl-4-2-fluoro-4-nitrophenyl, piperazine,1-ethyl-4-2-fluoro-4-nitrophenyl, 1-ethyl-4-2-fluoro-4-nitrophenyl piperazine
CCN1CCN(CC1)C2=C(C=C(C=C2)[N+](=O)[O-])F
C12H16FN3O2
XMJXGMZBNFICRS-UHFFFAOYSA-N
2947839

Specifications

1-Ethyl-4-(2-fluoro-4-nitrophenyl)piperazine
694501-34-3
15470780
XMJXGMZBNFICRS-UHFFFAOYSA-N
253.277
253.27
97%
1g
C12H16FN3O2
1-ethyl-4-2-fluoro-4-nitrophenyl piperazine, 1-ethyl-4-2-fluoro-4-nitro-phenyl-piperazine, piperazine, 1-ethyl-4-2-fluoro-4-nitrophenyl, piperazine,1-ethyl-4-2-fluoro-4-nitrophenyl, 1-ethyl-4-2-fluoro-4-nitrophenyl piperazine
CCN1CCN(CC1)C2=C(C=C(C=C2)[N+](=O)[O-])F
2947839
Solid
Videos
SDS