missing translation for 'onlineSavingsMsg'
Få mere at vide

1-Ethyl-3-piperidinone Hydrochloride, TRC

Artikelnummer. 30386270
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30386270 10 mg
30369201 100 mg
30377611 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30386270 Leverandør Toronto Research Chemicals Leverandørnr. TRCE93931810MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 41361-28-8
Molekylær formel C7H16ClNO2
Molekylvægt (g/mol) 181.66
Synonym 1-Ethyl-3-piperidone Hydrochloride
IUPAC navn 1-ethylpiperidin-3-one hydrochloride
SMIL Cl.O=C1CN(CC)CCC1

Specifications

Kemisk navn eller materiale 1-Ethyl-3-piperidinone Hydrochloride
Mængde 10 mg
InChI formel InChI=1S/C7H13NO.ClH/c1-2-8-5-3-4-7(9)6-8;/h2-6H2,1H3;1H
Formel vægt 163.08
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.