Learn More
1-Ethyl-3-methylimidazolium trifluoromethanesulfonate, 98+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
2 g
10 g
Description
Recently used in the preparation of hydrophobic, highly conductive ambient-temperature molten salts, and in the preparation of dual intercalating molten electrolyte batteries.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Recently used in the preparation of hydrophobic, highly conductive ambient-temperature molten salts, and in the preparation of dual intercalating molten electrolyte batteries.
Solubility
Soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture.
Recently used in the preparation of hydrophobic, highly conductive ambient-temperature molten salts, and in the preparation of dual intercalating molten electrolyte batteries.
Solubility
Soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture.
Specifications
Density | 1.387 |
Flash Point | None |
Refractive Index | 1.435 |
Quantity | 2 g |
Sensitivity | Hygroscopic |
Solubility Information | Soluble in water. |
Formula Weight | 260.24 |
Percent Purity | ≥98% |
Chemical Name or Material | 1-Ethyl-3-methylimidazolium trifluoromethanesulfonate |