Få mere at vide
1-Ethyl-2-[(methylamino)methyl]pyrrolidine, TRC
Artikelnummer. 17991185
Skift visning
Mængde:
25 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17991185 100 mg
17983195 50 mg
17961915 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 17991185 Leverandør Toronto Research Chemicals Leverandørnr. TRCE923645100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 60923-27-5
Molekylær formel C8 H18 N2
Molekylvægt (g/mol) 142.242
Synonym 1-Ethyl-N-methyl-2-pyrrolidinemethanamine
IUPAC navn 1-(1-ethylpyrrolidin-2-yl)-N-methyl-methanamine
SMIL CCN1CCCC1CNC
Tekniske data
Kemisk navn eller materiale 1-Ethyl-2-[(methylamino)methyl]pyrrolidine
Mængde 100 mg
InChI formel InChI=1S/C8H18N2/c1-3-10-6-4-5-8(10)7-9-2/h8-9H,3-7H2,1-2H3
Formel vægt 142.147