missing translation for 'onlineSavingsMsg'
Få mere at vide

1-Ethoxyethanimine-D3 Hydrochloride, TRC

Artikelnummer. 30371683
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30371683 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30371683 Leverandør Toronto Research Chemicals Leverandørnr. TRCE7492905G

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale 1-Ethoxyethanimine-D3 Hydrochloride
CAS 74929-87-6
Molekylær formel C4H6D3NO . HCl
Mængde 5 g
Synonym Ethyl Acetimidate HydrochlorideEthyl Acetoamidate HydrochlorideEthyl Acetocarboximidate HydrochlorideEthyl Acetoimidate HydrochlorideEthyl Ethanimidate HydrochlorideEthyl Ethanimidoate HydrochlorideEthyl Iminoacetate hydrochloride
InChI formel InChI=1S/C4H9NO.ClH/c1-3-6-4(2)5;/h5H,3H2,1-2H3;1H/i2D3;
SMIL Cl.[2H]C([2H])([2H])C(=N)OCC
IUPAC navn ethyl 2,2,2-trideuterioethanimidate;hydrochloride
Molekylvægt (g/mol) 126.6
Formel vægt 126.0639
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.