missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Eicosanethiol, 98%, Thermo Scientific Chemicals
311.50 DKK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
250 mg
1 g
Description
1-Eicosanethiol is used as an organic chemical synthesis intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Eicosanethiol is used as an organic chemical synthesis intermediate.
Notes
Air and moisture sensitive. Incompatible with heat.
1-Eicosanethiol is used as an organic chemical synthesis intermediate.
Notes
Air and moisture sensitive. Incompatible with heat.
Specifications
Melting Point | 38°C to 41°C |
Odor | Stench |
Linear Formula | CH3(CH2)19SH |
Quantity | 250 mg |
Sensitivity | Air sensitive |
Formula Weight | 314.61 |
Percent Purity | 98% |
Chemical Name or Material | 1-Eicosanethiol |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'