Få mere at vide
1-Dehydro Aldosterone, TRC
Artikelnummer. 30355007
Skift visning
Mængde:
0.5 mg
5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30355007 5 mg
30349652 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30355007 Leverandør Toronto Research Chemicals Leverandørnr. TRCD2291055MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 76959-24-5
Molekylær formel C21H26O5
Molekylvægt (g/mol) 358.43
Synonym (11beta)-11,21-Dihydroxy-3,20-dioxopregna-1,4-dien-18-al, delta1-Aldosterone, 11beta,21-Dihydroxy-3,20-dioxopregna-1,4-dien-18-al, 1-Dehydroaldosterone
SMIL C[C@]12C=CC(=O)C=C1CC[C@H]3[C@@H]4CC[C@H](C(=O)CO)[C@]4(C[C@H](O)[C@H]23)C=O
Tekniske data
Kemisk navn eller materiale 1-Dehydro Aldosterone
Mængde 5 mg
InChI formel InChI=1S/C21H28O5/c1-20-7-6-13(24)8-12(20)2-3-14-15-4-5-16(18(26)10-22)21(15,11-23)9-17(25)19(14)20/h6-8,14-17,19,22-23,25H,2-5,9-11H2,1H3/t14-,15+,16+,17-,19-,20-,21+/m0/s1
Formel vægt 358.178