Få mere at vide
1-Cyclopropylcarbonyl-piperazine-d8, TRC
Artikelnummer. 30361428
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30361428 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30361428 Leverandør Toronto Research Chemicals Leverandørnr. TRCC98668225MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1-Cyclopropylcarbonyl-piperazine-D8
CAS 2055113-57-8
Molekylær formel C8 D8 H6 N2 O
Mængde 25 mg
InChI formel InChI=1S/C8H14N2O/c11-8(7-1-2-7)10-5-3-9-4-6-10/h7,9H,1-6H2/i3D2,4D2,5D2,6D2
SMIL [2H]C1([2H])NC([2H])([2H])C([2H])([2H])N(C(=O)C2CC2)C1([2H])[2H]
IUPAC navn cyclopropyl-(2,2,3,3,5,5,6,6-octadeuteriopiperazin-1-yl)methanone
Molekylvægt (g/mol) 162.259
Formel vægt 162.161