missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
1-Cyclopropyl-7-8-difluoro-6-methoxy-4-oxo-1,4-dihydroquinoline-3-carboxylic Acid Ethyl Ester, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | 1-Cyclopropyl-7-8-difluoro-6-methoxy-4-oxo-1,4-dihydroquinoline-3-carboxylic Acid Ethyl Ester |
| CAS | 1329836-33-0 |
| Molekylær formel | C16 H15 F2 N O4 |
| Mængde | 1 mg |
| Synonym | 1-Cyclopropyl-7,8-difluoro-1,4-dihydro-6-methoxy-4-oxo-3-quinolinecarboxylic Acid Ethyl Ester |
| InChI formel | InChI=1S/C16H15F2NO4/c1-3-23-16(21)10-7-19(8-4-5-8)14-9(15(10)20)6-11(22-2)12(17)13(14)18/h6-8H,3-5H2,1-2H3 |
| SMIL | CCOC(=O)C1=CN(C2CC2)c3c(F)c(F)c(OC)cc3C1=O |
| IUPAC navn | ethyl 1-cyclopropyl-7,8-difluoro-6-methoxy-4-oxo-quinoline-3-carboxylate |
| Molekylvægt (g/mol) | 323.29 |
| Formel vægt | 323.0969 |
Produkttitel
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?