Få mere at vide
1-Cyclopropyl-4-oxo-7-(piperazin-1-yl)-1,4-dihydroquinoline-3-carboxylic Acid (Desfluoro Compound), Mikromol™
Artikelnummer. 17914016
Skift visning
Mængde:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17914016 25 mg
17932786 100 mg
30457636 4 x 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 17914016 Leverandør Mikromol Leverandørnr. MM0018.050025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
1-Cyclopropyl-4-oxo-7-(piperazin-1-yl)-1,4-dihydroquinoline-3-carboxylic Acid (Desfluoro Compound)
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Desfluoro Ciprofloxacin
CAS 93107-11-0
Analyt- eller komponentnavne Desfluoro Ciprofloxacin
Forsendelsestilstand Room Temperature
Molekylær formel C17 H19 N3 O3
Certificeringer/compliance ISO 9001
Synonym 1-Cyclopropyl-4-oxo-7-(piperazin-1-yl)-1,4-dihydroquinoline-3-carboxylic Acid, Ciprofloxacin Hydrochloride Imp. B (EP), Desfluoro Ciprofloxacin
InChI formel InChI=1S/C17H19N3O3/c21-16-13-4-3-12(19-7-5-18-6-8-19)9-15(13)20(11-1-2-11)10-14(16)17(22)23/h3-4,9-11,18H,1-2,5-8H2,(H,22,23)
SMIL OC(=O)C1=CN(C2CC2)c3cc(ccc3C1=O)N4CCNCC4
IUPAC navn 1-cyclopropyl-4-oxo-7-piperazin-1-ylquinoline-3-carboxylic acid
Molekylvægt (g/mol) 313.36
Mængde 25 mg
Formel vægt 313.357 g/mol