missing translation for 'onlineSavingsMsg'
Learn More

1-Cyclopentylpiperazine, TRC

Product Code. 30365632
Change view
Click to view available options
Mængde:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30365632 1 g
30355421 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30365632 Supplier Toronto Research Chemicals Supplier No. TRCC9890201G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21043-40-3
Molekylær formel C9H18N2
Molekylvægt (g/mol) 154.25
Synonym 1-Cyclopentylpiperazine, 4-Cyclopentylpiperazine, N-Cyclopentylpiperazine
IUPAC navn 1-cyclopentylpiperazine
SMIL C1CCC(C1)N2CCNCC2

Specifications

Kemisk navn eller materiale 1-Cyclopentylpiperazine
Mængde 1 g
InChI formel InChI=1S/C9H18N2/c1-2-4-9(3-1)11-7-5-10-6-8-11/h9-10H,1-8H2
Formel vægt 154.147
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.