Få mere at vide
1-Cyclopentylpiperazine, TRC
Artikelnummer. 30355421
Skift visning
Mængde:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30355421 5 g
30365632 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30355421 Leverandør Toronto Research Chemicals Leverandørnr. TRCC9890205G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 21043-40-3
Molekylær formel C9H18N2
Molekylvægt (g/mol) 154.25
Synonym 1-Cyclopentylpiperazine, 4-Cyclopentylpiperazine, N-Cyclopentylpiperazine
IUPAC navn 1-cyclopentylpiperazine
SMIL C1CCC(C1)N2CCNCC2
Tekniske data
Kemisk navn eller materiale 1-Cyclopentylpiperazine
Mængde 5 g
InChI formel InChI=1S/C9H18N2/c1-2-4-9(3-1)11-7-5-10-6-8-11/h9-10H,1-8H2
Formel vægt 154.147