Få mere at vide
1-Cyclohexyl-3-[[4-[2-[[(6-methylpyrazin-2-yl)carbonyl]amino]ethyl]phenyl]sulphonyl]urea, Mikromol™
Artikelnummer. 30052532
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30052532 100 mg
30049645 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30052532 Leverandør Mikromol Leverandørnr. MM0942.05
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
1-Cyclohexyl-3-[[4-[2-[[(6-methylpyrazin-2-yl)carbonyl]amino]ethyl]phenyl]sulphonyl]urea
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1-Cyclohexyl-3-[(4-[2-[((6-methylpyrazin-2-yl)carbonyl)amino]ethyl]phenyl)sulphonyl]urea
CAS 66375-96-0
Analyt- eller komponentnavne 1-Cyclohexyl-3-[(4-[2-[((6-methylpyrazin-2-yl)carbonyl)amino]ethyl]phenyl)sulphonyl]urea
Forsendelsestilstand Room Temperature
Molekylær formel C21 H27 N5 O4 S
Certificeringer/compliance ISO 9001
Synonym Glipizide Imp. E (EP), Glipizide USP Related Compound C, Glipizide USP RC C, 1-Cyclohexyl-3-[[4-[2-[[(6-methylpyrazin-2-yl)carbonyl]amino]ethyl]phenyl]sulphonyl]urea, Glipizide Related Compound C, Glipizide Impurity E
InChI formel InChI=1S/C21H27N5O4S/c1-15-13-22-14-19(24-15)20(27)23-12-11-16-7-9-18(10-8-16)31(29,30)26-21(28)25-17-5-3-2-4-6-17/h7-10,13-14,17H,2-6,11-12H2,1H3,(H,23,27)(H2,25,26,28)
SMIL Cc1cncc(n1)C(=O)NCCc2ccc(cc2)S(=O)(=O)NC(=O)NC3CCCCC3
IUPAC navn N-[2-[4-(cyclohexylcarbamoylsulfamoyl)phenyl]ethyl]-6-methylpyrazine-2-carboxamide
Molekylvægt (g/mol) 445.54
Mængde 100 mg
Formel vægt 445.1784 g/mol