Få mere at vide
1-Cyclohexyl-1-(4-benzyloxycyclohexyl)-2-(2-pyridinyl)ethanol-d11(Mixture of Diastereomers), TRC
Artikelnummer. 30360693
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30360693 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30360693 Leverandør Toronto Research Chemicals Leverandørnr. TRCC98791210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1-(Cyclohexyl-d11)-1-(4-benzyloxycyclohexyl)-2-(2-pyridinyl)
CAS 1189891-05-1
Molekylær formel C26H24D11NO2
Mængde 10 mg
Synonym 1-Cyclohexyl-1-(4-Benzyloxycyclohexyl)-2-(2-Pyridinyl)Ethanol-D11
InChI formel InChI=1S/C26H35NO2/c28-26(22-11-5-2-6-12-22,19-24-13-7-8-18-27-24)23-14-16-25(17-15-23)29-20-21-9-3-1-4-10-21/h1,3-4,7-10,13,18,22-23,25,28H,2,5-6,11-12,14-17,19-20H2/i2D2,5D2,6D2,11D2,12D2,22D
SMIL [2H]C1([2H])C([2H])([2H])C([2H])([2H])C([2H])(C(O)(Cc2ccccn2)C3CCC(CC3)OCc4ccccc4)C([2H])([2H])C1([2H])[2H]
IUPAC navn 1-(4-phenylmethoxycyclohexyl)-2-pyridin-2-yl-1-(1,2,2,3,3,4,4,5,5,6,6-undecadeuteriocyclohexyl)ethanol
Molekylvægt (g/mol) 404.63
Formel vægt 404.3358