missing translation for 'onlineSavingsMsg'
Learn More

1-Cyanomethyl-4,5-dicyano-2-(2-pyridyl)imidazole, 97%, Thermo Scientific™

Catalog Number p-7035881
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
1347814-89-4
C12H6N6
234.222
MFCD20265178
CKIOVYFZWSZUDP-UHFFFAOYSA-N
1-cyanomethyl-4,5-dicyano-2-2-pyridyl imidazole, 1-cyanomethyl-2-pyridin-2-yl imidazole-4,5-dicarbonitrile
73995634
1-(cyanomethyl)-2-pyridin-2-ylimidazole-4,5-dicarbonitrile
C1=CC=NC(=C1)C2=NC(=C(N2CC#N)C#N)C#N
This item is not returnable. View return policy
1347814-89-4
C12H6N6
234.222
MFCD20265178
CKIOVYFZWSZUDP-UHFFFAOYSA-N
1-cyanomethyl-4,5-dicyano-2-2-pyridyl imidazole, 1-cyanomethyl-2-pyridin-2-yl imidazole-4,5-dicarbonitrile
73995634
1-(cyanomethyl)-2-pyridin-2-ylimidazole-4,5-dicarbonitrile
C1=CC=NC(=C1)C2=NC(=C(N2CC#N)C#N)C#N

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
UN Number UN3439
Formula Weight 234.22
Percent Purity 97%
Chemical Name or Material 1-Cyanomethyl-4,5-dicyano-2-(2-pyridyl)imidazole
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'