Få mere at vide
1-Chloropropane-d7, CDN
Artikelnummer. 30325576
Skift visning
Mængde:
0.1 g
0.25 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30325576 0.25 g -
30316337 0.1 g 0.1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30325576 Leverandør CDN Leverandørnr. CDND64770.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1-Chloropropane-D7
CAS 761374-88-3
Molekylær formel C3 D7 Cl
Mængde 0.25 g
Synonym Propane-1,1,1,2,2,3,3-d7, 3-chloro- (9 CI), 3-Chloropropane-1,1,1,2,2,3,3-d7, 1-Chloropropane-d7, 1-Chloropropane-D7
InChI formel InChI=1 S/C3H7Cl/c1-2-3-4/h2-3H2,1H3/i1D3,2D2,3D2
SMIL [2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])Cl
IUPAC navn 1-chloro-1,1,2,2,3,3,3-heptadeuteriopropane
Molekylvægt (g/mol) 85.5838
Formel vægt 85.0676 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity