missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Chloromethyl-1H-1,2,4-triazole, hydrochloride, 98%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Specifications
Infrared Spectrum | Authentic |
Assay Percent Range | 97.5% min. (Argentometry) |
Packaging | Glass Bottle |
Quantity | 1 g |
Solubility Information | Solubility in water: soluble. Other solubilities: soluble in ethanol |
Formula Weight | 154 |
Percent Purity | 98% |
Chemical Name or Material | 1-Chloromethyl-1H-1,2,4-triazole, hydrochloride |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'