missing translation for 'onlineSavingsMsg'
Learn More

1-Chloromethyl-1H-1,2,4-triazole hydrochloride, 96%, Thermo Scientific Chemicals

Catalog Number p-7032391
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
84387-61-1
C3H4ClN3
117.54
MFCD00964947
PXNMFQMREJNQQT-UHFFFAOYSA-N
3743882
ClCN1C=NC=N1
This item is not returnable. View return policy
84387-61-1
C3H4ClN3
117.54
MFCD00964947
PXNMFQMREJNQQT-UHFFFAOYSA-N
3743882
ClCN1C=NC=N1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 138°C to 145°C
Quantity 250 mg
Sensitivity Hygroscopic
Synonym 1-chloromethyl-1h-1,2,4-triazole, 1-chloromethyl-1,2,4-triazole, 1h-1,2,4-triazole,1-chloromethyl, 1h-1,2,4-triazole, 1-chloromethyl, 1-chloro-methyl-1h-1,2,4-triazole
IUPAC Name 1-(chloromethyl)-1,2,4-triazole
Formula Weight 154
Percent Purity 96%
Chemical Name or Material 1-Chloromethyl-1H-1,2,4-triazole hydrochloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'