Få mere at vide
1-Chlorobutane-d9, CDN
Artikelnummer. 30328723
Skift visning
Mængde:
0.25 g
0.5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30328723 0.5 g
30314990 0.25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30328723 Leverandør CDN Leverandørnr. CDND10850.5G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1-Chlorobutane-D9
CAS 175540-76-8
Molekylær formel C4 D9 Cl
Mængde 0.5 g
Synonym Butane-1,1,1,2,2,3,3,4,4-d9, 4-chloro- (9 CI), 4-Chlorobutane-1,1,1,2,2,3,3,4,4-d9, 1-Chlorobutane-d9, 1-Chlorobutane-D9
InChI formel InChI=1 S/C4H9Cl/c1-2-3-4-5/h2-4H2,1H3/i1D3,2D2,3D2,4D2
SMIL [2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])Cl
IUPAC navn 1-chloro-1,1,2,2,3,3,4,4,4-nonadeuteriobutane
Molekylvægt (g/mol) 101.6227
Formel vægt 101.0958 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity