Få mere at vide
1-Chlorobenzotriazole, TRC
Artikelnummer. 30135984
Skift visning
Mængde:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30135984 10 g
30007149 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30135984 Leverandør Toronto Research Chemicals Leverandørnr. TRCC36453510G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 21050-95-3
Molekylær formel C6 H4 Cl N3
Molekylvægt (g/mol) 153.57
Synonym 1H-Benzotriazole, 1-chloro- (8CI, 9CI, ACI), 1-Chloro-1H-benzotriazole (ACI), 1-Chloro-1,2,3-benzotriazole, 1-Chloro-1H-1,2,3-benzotriazole, 1-Chlorobenzotriazole, N-Chlorobenzotriazole, NSC 186037
IUPAC navn 1-chlorobenzotriazole
SMIL Cln1nnc2ccccc12
Tekniske data
Kemisk navn eller materiale 1-Chlorobenzotriazole
Mængde 10 g
InChI formel InChI=1S/C6H4ClN3/c7-10-6-4-2-1-3-5(6)8-9-10/h1-4H
Formel vægt 153.0094