Få mere at vide
1-Chloroadamantane-d15, CDN
Artikelnummer. 30319069
Skift visning
Mængde:
0.01 g
0.05 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30319069 0.05 g
30318609 0.01 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30319069 Leverandør CDN Leverandørnr. CDND53030.05G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1-Chloroadamantane-D15
CAS 352431-55-1
Molekylær formel C10 D15 Cl
Mængde 0.05 g
Synonym Tricyclo[3.3.1.13,7 ]decane-1,2,2,3,4,4,5,6,6,8,8,9,9,10,10-d15, 7-chloro- (9 CI), -Chlorotricyclo[3.3.1.13,7 ]decane-1,2,2,3,4,4,5,6,6,8,8,9,9,10,10-d15, 1-Adamantyl-d15 chloride, 1-Chloroadamantane-d15, 1-Chloroadamantane-D15, 1-Chlorotricyclo[3.3.1.13,7 ]decane-d15
InChI formel InChI=1 S/C10H15Cl/c11-10-4-7-1-8(5-10)3-9(2-7)6-10/h7-9 H,1-6H2/i1D2,2D2,3D2,4D2,5D2,6D2,7 D,8 D,9 D
SMIL [2 H]C1([2 H])C2([2 H])C([2 H])([2 H])C3([2 H])C([2 H])([2 H])C1([2 H])C([2 H])([2 H])C(Cl)(C3([2 H])[2 H])C2([2 H])[2 H]
IUPAC navn 1-chloro-2,2,3,4,4,5,6,6,7,8,8,9,9,10,10-pentadecadeuterioadamantane
Molekylvægt (g/mol) 185.7715
Formel vægt 185.1804 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity