Få mere at vide
1-Chloro Dihydro Dutasteride, TRC
Artikelnummer. 30004497
Skift visning
Mængde:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30004497 50 mg
30011841 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30004497 Leverandør Toronto Research Chemicals Leverandørnr. TRCC36547550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1365545-42-1
Molekylær formel C27 H31 Cl F6 N2 O2
Molekylvægt (g/mol) 564.99
Synonym Dutasteride EP Impurity F, (4S,4aR,4bS,6aS,7S,9aS,9bS,11aR)-N-(2,5-Bis(trifluoromethyl)phenyl)-4-chloro-4a,6a-dimethyl-2-oxohexadecahydro-1H-indeno[5,4-f]quinoline-7-carboxamide, N-[2,5-Bis(trifluoromethyl)phenyl]-1α-chloro-3-oxo-4-aza-5α-androstane-17β-carboxamide, Dutasteride Imp F, Chlorodutasteride (USP)
IUPAC navn (1S,3aS,3bS,5aR,9S,9aR,9bS,11aS)-N-[2,5-bis(trifluoromethyl)phenyl]-9-chloro-9a,11a-dimethyl-7-oxo-1,2,3,3a,3b,4,5,5a,6,8,9,9b,10,11-tetradecahydroindeno[5,4-f]quinoline-1-carboxamide
SMIL C[C@]12CC[C@H]3[C@@H](CC[C@H]4NC(=O)C[C@H](Cl)[C@]34C)[C@@H]1CC[C@@H]2C(=O)Nc5cc(ccc5C(F)(F)F)C(F)(F)F
Tekniske data
Kemisk navn eller materiale 1-Chloro Dihydro Dutasteride
Mængde 50 mg
InChI formel InChI=1S/C27H31ClF6N2O2/c1-24-10-9-16-14(4-8-21-25(16,2)20(28)12-22(37)36-21)15(24)6-7-18(24)23(38)35-19-11-13(26(29,30)31)3-5-17(19)27(32,33)34/h3,5,11,14-16,18,20-21H,4,6-10,12H2,1-2H3,(H,35,38)(H,36,37)/t14-,15-,16-,18+,20-,21+,24-,25-/m0/s1
Formel vægt 564.1978