missing translation for 'onlineSavingsMsg'
Learn More

1-Chloro-2,5-bis(trifluoromethyl)benzene, 98%, Thermo Scientific™

Catalog Number p-7055660
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
328-91-6
C8H3ClF6
248.552
MFCD00632388
IDNVTCBYGYOPKJ-UHFFFAOYSA-N
2,5-bis trifluoromethyl chlorobenzene, 2-chloro-1,4-bis trifluoromethyl benzene, 1-chloro-2,5-bis trifluoromethyl benzene, 1,4-bis trifluoromethyl-2-chlorobenzene, 2,5-ditrifluoromethyl-chlorobenzene, 2-chlor-1,4-bis-trifluormethyl-benzol, 2-chloro-1,4-bis trifluoromethyl benzene #, benzene, 1-chloro-2,5-bis trifluoromethyl, benzene, 2-chloro-1,4-bis trifluoromethyl
619189
2-chloro-1,4-bis(trifluoromethyl)benzene
C1=CC(=C(C=C1C(F)(F)F)Cl)C(F)(F)F
This item is not returnable. View return policy
328-91-6
C8H3ClF6
248.552
MFCD00632388
IDNVTCBYGYOPKJ-UHFFFAOYSA-N
2,5-bis trifluoromethyl chlorobenzene, 2-chloro-1,4-bis trifluoromethyl benzene, 1-chloro-2,5-bis trifluoromethyl benzene, 1,4-bis trifluoromethyl-2-chlorobenzene, 2,5-ditrifluoromethyl-chlorobenzene, 2-chlor-1,4-bis-trifluormethyl-benzol, 2-chloro-1,4-bis trifluoromethyl benzene #, benzene, 1-chloro-2,5-bis trifluoromethyl, benzene, 2-chloro-1,4-bis trifluoromethyl
619189
2-chloro-1,4-bis(trifluoromethyl)benzene
C1=CC(=C(C=C1C(F)(F)F)Cl)C(F)(F)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It can be used as a synthetic building block.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY
Density 1.504 g/mL
Boiling Point 147°C to 149°C
Refractive Index 1.4135
Quantity 1 g
Beilstein 1978018
Solubility Information Slightly soluble in water.
Formula Weight 248.55
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1-Chloro-2,5-bis(trifluoromethyl)benzene
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'