Få mere at vide
1-C-[3-[[5-(4-Fluorophenyl)-2-thienyl]methyl]-4-methylphenyl]-D-glucose, TRC
Artikelnummer. 30179651
Skift visning
Mængde:
50 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30179651 500 mg
30179203 50 mg
30179545 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30179651 Leverandør Toronto Research Chemicals Leverandørnr. TRCF828210500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1809403-04-0
Molekylær formel C24 H25 F O6 S
Molekylvægt (g/mol) 460.52
Synonym (2R,3S,4R,5R)-1-(3-((5-(4-Fluorophenyl)thiophen-2-yl)methyl)-4-methylphenyl)-2,3,4,5,6-pentahydroxyhexan-1-one
IUPAC navn (2R,3S,4R,5R)-1-[3-[[5-(4-fluorophenyl)thiophen-2-yl]methyl]-4-methylphenyl]-2,3,4,5,6-pentahydroxyhexan-1-one
SMIL Cc1ccc(cc1Cc2ccc(s2)c3ccc(F)cc3)C(=O)[C@H](O)[C@@H](O)[C@H](O)[C@H](O)CO
Tekniske data
Kemisk navn eller materiale 1-C-(3-((5-(4-Fluorophenyl)-2-thienyl)methyl)-4-methylphenyl)-D-glucose
Mængde 500 mg
InChI formel InChI=1S/C24H25FO6S/c1-13-2-3-15(21(28)23(30)24(31)22(29)19(27)12-26)10-16(13)11-18-8-9-20(32-18)14-4-6-17(25)7-5-14/h2-10,19,22-24,26-27,29-31H,11-12H2,1H3/t19-,22-,23+,24+/m1/s1
Formel vægt 460.1356