Få mere at vide
1-Butanamine-d9, TRC
Artikelnummer. 30342961
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
30342961 100 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30342961 Leverandør Toronto Research Chemicals Leverandørnr. TRCB690022100MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1-Butanamine-d9
CAS 776285-22-4
Molekylær formel C4H2D9N
Mængde 100 mg
Synonym n-Butyl-d9-amine, 1-Aminobutane-d9, 1-Butylamine-d9, Mono-n-butylamine-d9, Monobutylamine-d9, NSC 8029-d9, Norvalamine-d9, n-Butylamine-d9
InChI formel InChI=1S/C4H11N/c1-2-3-4-5/h2-5H2,1H3/i1D3,2D2,3D2,4D2
SMIL NC([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])[2H]
IUPAC navn butan-d9-1-amine
Molekylvægt (g/mol) 82.19
Formel vægt 82.15