Få mere at vide
1-Bromoundecane-11,11,11-d3, CDN
Artikelnummer. 30324120
Skift visning
Mængde:
0.1 g
0.25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30324120 0.1 g
30314038 0.25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30324120 Leverandør CDN Leverandørnr. CDND63570.1G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1-Bromoundecane-11,11,11-D3
CAS 1219805-63-6
Molekylær formel C11 D3 H20 Br
Mængde 0.1 g
Synonym 11-Bromoundecane-1,1,1-d3, 1-Bromoundecane-11,11,11-d3, 1-Bromoundecane-11,11,11-D3, 1-Undecyl-11,11,11-d3 bromide
InChI formel InChI=1 S/C11H23Br/c1-2-3-4-5-6-7-8-9-10-11-12/h2-11H2,1H3/i1D3
SMIL [2 H]C([2 H])([2 H])CCCCCCCCCCBr
IUPAC navn 11-bromo-1,1,1-trideuterioundecane
Molekylvægt (g/mol) 238.2228
Formel vægt 237.1171 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity